Learn More
IL-12 (p70) Rat anti-Human, PE, Clone: 20C2, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 559325
Description
The 20C2 antibody reacts with the biologically active form of human IL-12 p70 heterodimer, but not with the p40 monomer or homodimer. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development or reported in the literature only.
Intracellular Staining

Specifications
IL-12 (p70) | |
Monoclonal | |
PE | |
Rat | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
20C2 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.