Learn More
anti-IL-12 R beta 1, APC, Clone: 69310, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB839A025
 View more versions of this product
Catalog No. FAB839A025
Add to cart
This item is not returnable. View return policy
Description
Description
IL-12 R beta 1 Monoclonal antibody specifically detects IL-12 R beta 1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-12 R beta 1
69310
Flow Cytometry 10 uL/10^6 cells
P42701
CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL12R beta 1, IL-12R subunit beta-1, IL12RB, IL12RB1, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454
Mouse
IgG1
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human IL-12 R beta 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-12 R beta 1 is observed.
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
IL12RB1
Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 1 Cys24-Glu540 Accession # P42701
Protein A or G purified from ascites
RUO
Primary
3594
Human