Learn More
anti-IL-12 R beta 1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP132449
Catalog No. NBP132449
Add to cart
Description
Description
IL-12 R beta 1 Polyclonal antibody specifically detects IL-12 R beta 1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IL-12 R beta 1
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
IL12RB1
Recombinant protein encompassing a sequence within the center region of human IL12 Receptor beta1. The exact sequence is proprietary.
73 kDa
100 ul
Cell Cycle and Replication
Primary
3594
Human
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL-12R subunit beta-1, IL12RB, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.