Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL12RB1 Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA124807

Catalog No. PA124807


Add to cart

Description

Description

PA1-24807 detects IL12RB1 from human samples. PA1-24807 has been successfully used in Western blot procedures. The PA1-24807 immunogen is recombit, full length human protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

IL12RB1
Polyclonal
Unconjugated
IL12RB1
Liquid
IL12RB1
Recombit, full length human protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
0.1 mg/mL
PBS with no preservative
P42701
IL12RB1, interleukin 12 receptor, beta 1, CD212, IL-12R-BETA1, IL12RB, MGC34454, IL-12 receptor beta component, interleukin-12 receptor beta-1 chain
Goat
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3594
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit