Learn More
IL-12R-beta-1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061089
Catalog No. 06-108-9MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect IL-12R-beta-1 using this Anti-IL-12R-beta-1 Antibody validated for use in western blot, immunohistochemistry.
Specifications
Specifications
IL-12R-beta-1
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human
Immunohistochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
IL-12RB1; CD212; IL-12R-BETA1; IL12RB; IL12RB1; IL12R
KLH-conjugated linear peptide corresponding to human IL-12R-beta-1 at the cytoplasmic domain.
100μg
Primary
NP_005526