Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL-13, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142776

Catalog No. NBP142776


Add to cart

Description

Description

IL-13 Polyclonal antibody specifically detects IL-13 in Porcine, Bovine, Sheep samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

IL-13
LYOPH
Western Blot 1.5-2 ug/ml, ELISA 1:11000
Purified
IL13
This protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant protein raised in yeast, corresponding to mature porcine IL-13 protein.
Protein A purified
RUO
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific for swine IL-13 protein. A BLAST analysis was used to suggest reactivity with IL-13 from swine sources based on 100% identity with the immunizing sequence. The following homologies may indicate chances of cross-reactivity when amino acid similarity and not just exact sequence is accounted: 89-90% homology to IL-13 from sheep, dolphin, beluga, and bovine, 83-88% to human and certain monkey IL-13, 80-82% to dusky titi, macaque, baboon, mangabey, dog, llama, camel, and galago, 75-78% to horse, and 67-72% to rat, mouse, and gerbil. Cross-reactivity with IL-13 from other sources has not been determined.
Bovine, Porcine, Sheep
ELISA, Western Blot
Unconjugated
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
Rabbit
IgG
0.1 mg
Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary
3596
Reconstitute with 100 ul deionized water.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only