Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13 R alpha 2 Mouse anti-Human, Clone: 83834, R&D Systems™

Mouse Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen IL-13R alpha 2
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-13 R alpha 2 Cys22-Leu342 Accession # Q14627
Clone 83834
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB614SP
View Documents
R&D Systems
MAB614SP
25 ug
Each for $114.50
Add to cart
 
MAB614100
View Documents
R&D Systems
MAB614100
100 ug
Each for $377.00
Add to cart
 
MAB614500
View Documents
R&D Systems
MAB614500
500 ug
Each for $839.00
Add to cart
 
Description

Description

IL-13 R alpha 2 Monoclonal antibody MAB614 Specifically detects IL-13 R alpha 2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

IL-13R alpha 2
83834
Unconjugated
Q14627
Monoclonal
Primary
Detects human IL-13 R alpha 2 in direct ELISAs and Western blots. In Western blots, this antibody shows approximately 5% cross-reactivity with recombinant human (rh) IL-4 R and rhIL-9 R and no cross-reactivity with rhIL-5 R alpha , rhIL-5 R beta , rhIL-13 R alpha 1, or recombinant mouse IL-13 R alpha 2.
Protein A or G purified from hybridoma culture supernatant
Mouse myeloma cell line NS0-derived recombinant human IL-13 R alpha 2 Cys22-Leu342 Accession # Q14627
Western Blot
Purified
Mouse
IgG1
3598
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit