Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL13RA1 Mouse anti-Human, Clone: GM1E7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA117751

Catalog No. 01-670-164


Add to cart

Description

Description

This antibody recognizes native IL13R protein expressed in transfected mammalian cells.

Interleukin 13 (IL-13) is a T cell derived cytokine involved in the regulation of inflammatory and im-mune responses. IL-13Ra1 together with IL-4Ra forms a functional receptor for both IL-4 and IL-13, which is why these two cytokines share many of their biological activities. The receptor is found on human B cells, monocytes and endothelial cells. However, no functional receptor is expressed on T cells, which explains why IL-13, in contrast to IL-4, fails to induce TH2-cell differentiation.
Specifications

Specifications

IL13RA1
Monoclonal
2 mg/mL
PBS with no preservative; pH 7.2
P78552
CD213A1, IL-13Ra, NR4, CD213a1 antigen, CT19, IL-13 receptor subunit alpha-1, IL-13R subunit alpha-1, IL-13R-alpha-1, IL-13RA1, IL13 receptor alpha-1 chain, Interleukin 13 receptor
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3597
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
GM1E7
Unconjugated
IL13RA1
Liquid
IL13RA1
cDNA encoding human IL13-R alpha 1 (extracellular domain)
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit