Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13 Mouse anti-Human, PE, Clone: 32007, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523560

Catalog No. PIMA523560


Add to cart

Description

Description

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
Specifications

Specifications

IL-13
Monoclonal
PE
IL13
Liquid
IL13
Human IL-13
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
32007
Buffered saline solution with BSA and 0.09% sodium azide
P35225
ALRH, BHR1, IL-13, MGC116786, MGC116788, MGC116789, P600, interleukin-13
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3596
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit