Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-16 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3161
This item is not returnable.
View return policy
Description
IL-16 Monoclonal specifically detects IL-16 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| IL-16 | |
| Monoclonal | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells | |
| Q14005 | |
| IL16 | |
| E. coli-derived recombinant human IL-16 isoform 1 Met1203-Ser1332 Accession # Q14005 | |
| 100 μg | |
| Primary | |
| Detects human IL-16. | |
| Human | |
| Purified |
| Flow Cytometry | |
| 70720 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ16806, FLJ42735, FLJ44234, HsT19289, IL16, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCF, LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3603 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction