Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520670
Description
A suggested positive control is rat brain tissue lysate. PA5-20670 can be used with blocking peptide PEP-0788.
IL-16 Polyclonal antibody specifically detects IL-16 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
IL-16 | |
Polyclonal | |
Unconjugated | |
IL16 | |
FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116996, 16170, 3603 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O54824, Q14005 | |
IL16 | |
A 20 amino acid peptide near the amino terminus of human IL-16. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction