Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL17RB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141211

Catalog No. 01-675-771


Add to cart

Description

Description

Suggested positive control: Hela whole cell extract, HeLa cell lysate.

Recently, a number of cytokines belonging to the interleukin (IL)-17 family have been identified. These are IL-17B, IL-17C, and IL-17E. Like IL-17, IL-17B, C, and E are able to induce proinflammatory responses but don ft.t bind to the IL-17 receptor suggesting additional IL-17R related receptors might exist. IL-17E induces activation of NF-kB pathway and like IL-17 also induces production of IL-8. IL-17Rh1 mRNA is highly expressed in kidney and liver and at moderate level in small intestine, colon, and brain.
Specifications

Specifications

IL17RB
Polyclonal
Unconjugated
IL17RB
Liquid
IL17RB
Peptide corresponding to residues C(102) N Y T E A F Q T Q T R P S G G K(118) of human IL-17BR.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.1mg/mL BSA and 0.02% sodium azide
Q9NRM6
CRL4, EVI27, IL17BR, IL17RH1, MGC5245
Rabbit
IgG
200 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
55540
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit