Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17E/IL-25 Mouse anti-Human, Clone: 182203, R&D Systems™

Mouse Monoclonal Antibody

$125.00 - $839.00

Specifications

Antigen IL-17E/IL-25
Immunogen E. coli-derived recombinant human IL-17E Tyr33-Gly177 Accession # Q9H293
Clone 182203
Applications Western Blot, Flow Cytometry, CyTOF
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1258SP
View Documents
R&D Systems
MAB1258SP
25 ug
Each for $125.00
Add to cart
 
MAB1258100
View Documents
R&D Systems
MAB1258100
100 ug
Each for $377.00
Add to cart
 
MAB1258500
View Documents
R&D Systems
MAB1258500
500 ug
Each for $839.00
Add to cart
 
Description

Description

IL-17E/IL-25 Monoclonal antibody MAB1258 Specifically detects IL-17E/IL-25 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Western Blotting
Specifications

Specifications

IL-17E/IL-25
182203
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG1
64806
RUO
E. coli-derived recombinant human IL-17E Tyr33-Gly177 Accession # Q9H293
Western Blot, Flow Cytometry, CyTOF
Purified
Q9H293
Monoclonal
Primary
Detects human IL-17E in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse IL-17E and no cross-reactivity with recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, or rhIL-17F.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit