Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL-17F, Biotin, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP142749

Catalog No. NBP142749

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

IL-17F
LYOPH
Western Blot 1:2000-1:10000, ELISA 1:20000-1:100000, Immunohistochemistry 1:1000-1:5000, Immunohistochemistry-Paraffin 1:10-1:500
Affinity Purified
IL17F
This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant human IL17-F protein.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
In ELISA and other immunoreactive assays, this antibody will recognize both native and recombinant human IL17-F in cell supernatants and certain body fluids. A control of similarly diluted normal rabbit IgG is recommended.
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Cytokine ML-1, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24
Rabbit
IgG
0.1 mg
Cytokine Research
Primary
112744
Reconstitute with 50 ul of distilled water to a final concentration of 1.0 mg/ml.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only