Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL18BP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14118

Catalog No. PA14118


Add to cart

Description

Description

PA1-4118 detects Interleukin-18 binding protein in human samples. PA1-4118 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~40 kDa protein representing Interleukin-18 binding protein in human spleen lysate. The PA1-4118 immunogen is a synthetic peptide corresponding to residues S(44) T K D P C P S Q P P V (55) of human IL-18BP.

IL-18 is a cytokine and acts as an early signal in the development of the helper T cell (Th1) response. IL-18BP is a binding protein binds IL-18 and neutralizes the biological activity of IL-18 in vitro.
Specifications

Specifications

IL18BP
Polyclonal
Unconjugated
IL18BP
Liquid
IL18BP
Synthetic peptide corresponding to residues S(44) T K D P C P S Q P P V (55) of human IL-18BP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O95998
Interleukin-18 binding protein, IL18BPa, Tadekinig-alfa
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10068
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit