Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18 Mouse anti-Human, Mouse, Clone: 12E7.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 041585
Description
Anti-IL-18 Antibody, clone 12E7.1 is an antibody against IL-18 for use in immunohistochemistry, flow cytometry.Specifications
IL-18 | |
Monoclonal | |
Unconjugated | |
NP_001553 | |
Mouse | |
Ion Exchange | |
Primary | |
2-8°C for one year | |
IgM κ |
Flow Cytometry, Immunohistochemistry | |
12E7.1 | |
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide. | |
IL-18, IL18, IGIF, IL-18, IL1F4, IL-1g | |
KLH-conjugated linear peptide corresponding to human IL-18. | |
100 μg | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only