Learn More
anti-IL-18 R alpha/IL-1 R5, PE, Clone: 70625, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB840P025
 View more versions of this product
Catalog No. FAB840P025
Add to cart
This item is not returnable. View return policy
Description
Description
IL-18R alpha/IL-1 R5 Monoclonal antibody specifically detects IL-18R alpha/IL-1 R5 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
IL-18R alpha/IL-1 R5
70625
Flow Cytometry 10 uL/10^6 cells
Q13478
IL18R1
Mouse myeloma cell line NS0-derived recombinant human IL-18 R alpha/IL-1 R5 Glu20-Arg329 Accession # Q13478
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8809
Human
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
Mouse
IgG1
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human IL-18 R alpha/IL-1 R5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-1 R3, rhIL-1 R4, rhIL-1 R6, rhIL-1 R7, rhIL-1 R8, rhIL-1 R9, rhSIGIRR or recombinant mouse IL-18 R alpha is observed.