Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-19 Monoclonal Antibody (350105), Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA524093
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant humanIL-19 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-19 Monoclonal antibody specifically detects IL-19 in Mouse samples. It is validated for Flow Cytometry, Western Blot.Specifications
IL-19 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8CJ70 | |
IL19 | |
E. coli-derived recombinant mouse IL-19 Leu25-Ala176 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
350105 | |
Unconjugated | |
IL19 | |
il 19; IL-10C; IL19; IL-19; ILN; Interleukin; interleukin 19; Interleukin19; interleukin-19; MDA1; MDA-1; melanoma differentiation associated protein-like protein; melanoma differentiation-associated protein-like protein; NG.1; ZMDA1 | |
Rat | |
Protein A/G | |
RUO | |
329244 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction