Learn More
IL-2 Mouse anti-Human, Brilliant Violet 650, Clone: 5344.111, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563467
Catalog No. BDB563467
Add to cart
Description
Description
The 5344.111 antibody specifically binds to the multifunctional cytokine, human interleukin-2 (IL-2). IL-2 is produced by activated T cells and has multiple functions that can affect the growth, proliferation and differentiation of many different target cell types including T cells, B cells, NK cells, monocytes and macrophages. The immunogen used to generate the 5344.111 hybridoma was affinity-purified, recombinant human IL-2 protein.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon™ BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will bespillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
Specifications
IL-2
5344.111
IL2; Interleukin-2; T-cell growth factor; TCGF
Affinity Purified
Human IL-2 protein
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 650
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human