Learn More
IL-2 Mouse anti-Human, Clone: 297C 16G2, Invitrogen
Mouse Monoclonal Antibody
Manufacturer: Invitrogen AHC0622
Description
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
IL-2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.4 | |
P60568 | |
T-cell growth factor, TCGF, interleukin-2 | |
Mouse | |
IgG2b, kappa | |
500 μg | |
4° C | |
Primary | |
3558 |
ELISA, Immunohistochemistry | |
297C 16G2 | |
Unconjugated | |
IL2 | |
Liquid | |
IL2 | |
Purified recombit human IL-2 | |
Protein A | |
RUO | |
Antibody | |
Monoclonal | |
Human |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.