Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-2 Mouse anti-Human, FITC, Clone: 5334, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523580

Catalog No. PIMA523580


Add to cart

Description

Description

This antibody does not cross-react with Recombinant IL-2 from mouse, rat, pig, or cotton rat.

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications

Specifications

IL-2
Monoclonal
FITC
IL2
Liquid
IL2
E. coli-derived recombit human IL-2 Ala21-Thr153
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
5334
Buffered saline solution with BSA and 0.1% sodium azide
P60568
T-cell growth factor, TCGF, interleukin-2
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3558
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit