Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL17B Mouse anti-Human, APC, Clone: 174113, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523668

Catalog No. PIMA523668


Add to cart

Description

Description

In Western blots, approximately 25% cross-reactivity with recombit mouse IL-17B is observed and no cross-reactivity with recombit human (rh) IL-17, rhIL-17C, rhIL-17E or rhIL-17F is observed.

Specifications

Specifications

IL17B
Monoclonal
APC
IL17B
Liquid
IL17B
E. coli-derived recombit human IL-17B Gln21-Phe180
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
174113
Buffered saline solution with BSA and 0.09% sodium azide
Q9UHF5
IL17B, interleukin 17B, NIRF, ZCYTO7, cytokine Zcyto7, cytokine-like protein ZCYTO7, interleukin 20, interleukin-17 beta, interleukin-17B, interleukin-20, neuronal interleukin-17 related factor, neuronal interleukin-17-related factor
Mouse
IgG2B
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
27190
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit