Learn More
IL-21 Receptor Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061075
Catalog No. 06-107-5MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This Anti-IL-21 Receptor Antibody is validated for use in western blot, immunohistochemistry, FC for the detection of IL-21 Receptor.
Specifications
Specifications
IL-21 Receptor
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Human, Rat
Flow Cytometry, Immunohistochemistry, Western Blot
Purified rabbit polyclonal in PBS with 0.09% sodium azide.
IL21R; IL-21R; NILR
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human IL-21 Receptor.
100μg
Primary
NP_068570