Learn More
anti-IL-22, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP131191
Catalog No. NBP131191
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
IL-22
1.0mg/mL
Western Blot 1:500-1:3000, ELISA 1:1000-1:10000, Immunohistochemistry :100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
IL22
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human IL22. The exact sequence is proprietary.
20 kDa
100 ul
Cytokine Research, Innate Immunity
Primary
50616
Human
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-21, IL-22IL21, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 21, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (86%), Rat (82%), and Rhesus Monkey (96%). Not yet tested in any other species.