Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22 Receptor alpha Rat anti-Mouse, Clone: 496514, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524205

Catalog No. PIMA524205


Add to cart

Description

Description

In direct ELISAs, 25% cross-reactivity with recombit human (rh) IL-22 R alpha 1 is observed and no cross-reactivity with rhIL-20 R alpha, recombit mouse (rm) IL-20 R alpha, rhIL-22BP, or rmIL-22BP is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

A novel cytokine, designated IL-TIF for IL-10 related T cell-derived inducible factor and IL-22, was recently identified. The receptor for IL-22 (IL-22R, also termed CRF2-9 and IL-TIF-R1 chain) is a new member of the class II cytokine receptor family. IL-22R forms a complex with IL-10 receptor beta chain and mediates IL-22 signaling. IL-22 and its receptor activate JAK-STAT signaling pathway. IL22R is expressed in normal liver and kidney and their cell lines HepG2 and TK-10. A soluble form of IL-22 receptor, also termed IL-22 binding protein (IL-22BP) and IL-22 receptor-alpha 2 (IL-22RA2), was identified very recently. IL-22BP prevents binding of IL-22 to the functional cell surface IL-22R complex and neutralizes IL-22 activity. LPS induces IL-22 expression, which indicates the role of IL-22 in inflammatory response.
Specifications

Specifications

IL-22 Receptor alpha
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q80XZ4
CRF2-9, IL22R, IL22R1, IL-22 receptor subunit alpha-1, IL-22R-alpha-1, IL-22RA1, cytokine receptor class-II member 9, cytokine receptor family 2 member 9, interleukin-22 receptor subunit alpha-1, zcytoR11
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
230828
Flow Cytometry, Immunocytochemistry, Immunofluorescence
496514
Unconjugated
Il22ra1
Lyophilized
IL22RA1
Mouse myeloma cell line NS0-derived recombit mouse IL-22 R alpha 1 Thr18-Ala228
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit