Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-IL-22RA-1, Polyclonal, ™ Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ 061077/DEL

Catalog No. 50-171-948

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Interleukin 22 Receptor α 1 (IL-22RA-1) is a member of the type II cytokine receptor family and has recently been discovered to be produced by Th17 cells. IL-22RA-1 and IL-10RB form a complex that mediates IL-22 signaling via JAK/STAT pathways. Current studies have revealed that IL-22 targets cells of the digestive, skin, and respiratory organ systems and plays an important role in mucosal immunity. The IL-22 receptor (IL-22R) is expressed exclusively in these tissues, thereby allowing the cytokine to mediate epithelial innate immunity in response to a variety of pathogens. Although recent studies have shown the importance of IL-22 in host defense against Gram-negative bacterial organisms (in gut and lung), there is evidence that IL-22 also plays a role in autoimmune disease, such as psoriasis. IL-22 therefore, not unlike other cytokines, has complex pro-inflammatory, anti-inflammatory, and autoimmune effects.

Host Species: Rabbit
Cross Reactivity: Chimpanzee, Human, Mouse
Immunogen: KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human IL-22R, α 1 subunit.

Flow Cytometry, Immunohistochemistry, Western Blotting

Specifications

Specifications

IL-22RA-1
Affinity Purified
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human IL-22R, α 1 subunit.
Primary
Chimpanzee, Human, Murine
Unconjugated
Rabbit
100μg
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit