Learn More
IL-23 Rat anti-Mouse, PerCP-Cy5.5, Clone: O78-1208, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 564828
Description
The O78-1208 monoclonal antibody specifically binds to the mouse Interleukin-23 Receptor (IL-23R) subunit that is encoded by the il23r gene. The IL-23R subunit is a type I transmembrane glycoprotein and member of the hemopoietin receptor superfamily. The mouse IL-23 Receptor complex is comprised of IL-23R and IL-12 receptor beta 1 (IL-12 Rβ1) subunit s. The IL-23R complex can bind IL-23, a cytokine that plays roles in innate and adaptive immunity as well as in autoimmune diseases, eg, by the generation and maintenance of Th17 cells. Mouse IL-23R is expressed by activated/memory CD4+ T cells, Th1, Th2 and Th17 cells, γδ T cells, dendritic cells and macrophages as determined by IL-23R mRNA expression and IL-23R-GFP reporter mouse studies. The IL-23-bound IL-23R complex transduces an intracellular signal pathway mediated by a Jak-STAT signaling cascade.
Specifications
IL-23 | |
Monoclonal | |
0.2mg/mL | |
Il23r; IL-23R; Interleukin 23 receptor | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
O78-1208 | |
PerCP-Cyanine5.5 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse IL-23 Receptor Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.