Learn More
anti-IL-23 R, PerCP, Clone: 753317, R&D Systems™
Manufacturer:  R&D Systems FAB16861C025
 View more versions of this product
Catalog No. FAB16861C02
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
IL-23 R
753317
Supplied in a saline solution containing BSA and Sodium Azide.
Purified
Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R, Gly24-Asp372
Protein A or G purified from hybridoma culture supernatant
RUO
Monoclonal
Detects endogenous mouse IL-23 R by flow cytometry.
Flow Cytometry
PerCP
Q8K4B4
Rat
IgG1
25 Tests
Primary
209590
Murine