Learn More
anti-IL-23 R, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP128827
Catalog No. NBP128827
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
IL-23 R
1.0 mg/mL
Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:25
Q5VWK5
IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Goat
IgG
0.05 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
IL-23 Receptor
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS, with 0.1% BSA with 0.1% Sodium Azide
Purified
IL23R
Synthetic peptide CQPRKLHFYKNGIKERFQITRINKTTARL corresponding to amino acids 59-87 within the N-terminal region of human IL-23 receptor.
IgG purified
RUO
Primary
149233
Human, Mouse