Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-IL-23R, cytoplasmic, Polyclonal, ™ Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ 061331/DEL

Catalog No. 50-171-989

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Interleukin 23 receptor (IL-23R) is a type I cytokine receptor that is a subunit of the receptor for IL23A/IL23. IL-23R pairs with the receptor molecule IL12RB1/IL12Rβ1, and both are required for IL23A signaling. IL-23R associates constitutively with Janus kinase 2 (JAK2), and also binds to transcription activator STAT3.

Host Species: Rabbit
Cross Reactivity: Human, Mouse
Immunogen: KLH-conjugated linear peptide corresponding to human IL-23R at and around the cytoplasmic domain.

ELISA, Flow Cytometry, Immunohistochemistry, Western Blotting

Specifications

Specifications

IL-23R, cytoplasmic
Affinity Purified
KLH-conjugated linear peptide corresponding to human IL-23R at and around the cytoplasmic domain.
Primary
Human, Murine
Unconjugated
Rabbit
100μg
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit