Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-25 Mouse anti-Human, FITC, Clone: 182203, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523602

Catalog No. PIMA523602

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.
Specifications

Specifications

IL-25
Monoclonal
FITC
IL25
Liquid
IL25
E. coli-derived recombit human IL-17E/IL-25 Tyr33-Gly177
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
182203
Buffered saline solution with BSA and 0.09% sodium azide
Q9H293
IL25, interleukin 25, IL17E, interleukin 17E, interleukin-17E, interleukin-25
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
64806
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit