Learn More
IL-25 (IL-17E) Rabbit anti-Human, Mouse, Chimpanzee, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061080
Catalog No. 06-108-0MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect IL-25 (IL-17E) using this Anti-IL-25 (IL-17E) Antibody, N-terminus validated for use in immunohistochemistry, ELISA, flow cytometry.
Specifications
Specifications
IL-25 (IL-17E)
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Chimpanzee, Human, Murine
ELISA, Flow Cytometry, Immunohistochemistry
Purified rabbit polyclonal in PBS with 0.05% sodium azide.
IL25; IL-17E; IL-25; IL17E; Interleukin-17E
KLH-conjugated linear peptide corresponding to human IL-25 at and around the N-terminus.
100μg
Primary
NP_073626