Learn More
IL-26 Rabbit anti-Human, Monkey, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061081
Catalog No. 06-108-1MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect IL-26 using this Anti-IL-26 Antibody validated for use in western blot, flow cytometry.
Specifications
Specifications
IL-26
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Human, Monkey
Flow Cytometry, Western Blot
Purified rabbit polyclonal in PBS with 0.09% sodium azide.
IL26; AK155; IL-26
KLH-conjugated linear peptide corresponding to human IL-26 near the C-terminus.
100μg
Primary
NP_060872