Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-27 Monoclonal Antibody (307426), APC, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA523573
Description
IL-27 Monoclonal antibody specifically detects IL-27 in Human samples. It is validated for Flow Cytometry.Specifications
IL-27 | |
Monoclonal | |
APC | |
Ebi3 | |
cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; Il27; IL-27; IL27 p28; IL-27 p28 subunit; IL27 subunit; IL-27 subunit alpha; IL-27 subunit beta; Il27a; IL-27A; IL-27-A; IL27-A; Il27b; IL-27B; IL27p28; IL-27p28; Il30; IL35 subunit; ILN; Interleukin; interleukin 27; interleukin 30; interleukin-27 subunit alpha; Interleukin-27 subunit beta; interleukin-30; MGC71873; M-IL-27 (p28); p28; RGD1561420 | |
Mouse | |
Protein A/G | |
RUO | |
10148, 246778 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
307426 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
Q14213, Q8NEV9 | |
Ebi3, IL27 | |
Mouse myeloma cell line NS0-derived recombinant human IL-27 Arg21-Lys229 of EBI-3 and Phe29-Pro243 of p28 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction