Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL17D, PE, clone: 312724, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523606

Catalog No. PIMA523606


Add to cart

Description

Description

The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. The treatment of endothelial cells with this cytokine has been shown to stimulate the production of other cytokines including IL6, IL8 and CSF2/ GM-CSF. The increased expression of IL8 induced by this cytokine was found to be NF-kappa B-dependent.

In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-17D, recombinant mouse (rm) IL-17, rmIL-17B, rmIL-17C, rmIL-17E, or rmIL-17F is observed.
Specifications

Specifications

IL17D
Monoclonal
PE
Ebi3
Liquid
EBI3, IL17D, IL27
E. coli-derived recombinant mouse IL-17D Ala25-Arg205
Protein A/G
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
312724
Buffered saline solution with BSA and 0.1% sodium azide
O35228, Q8K3I6
IL-3R
Rat
IgG2A
100 Tests
Store at 4°C, store in the dark
Primary
239114, 246779, 50498
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit