Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IL-27 R alpha/WSX-1/TCCR, Alexa Fluor 700, Clone: 263503, R&D Systems
Manufacturer: R&D Systems FAB21091N100
Specifications
IL-27 R alpha/WSX-1/TCCR | |
263503 | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
Purified | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-27 Rα/WSX‑1/TCCR, Gly29-Lys510 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Monoclonal | |
Detects mouse IL-27 Rα/WSX‑1/TCCR in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse gp130 or recombinant human IL-27 Rα is observed. |
Flow Cytometry | |
Alexa Fluor 700 | |
O70394 | |
Rat | |
IgG2b | |
100 Tests | |
Primary | |
50931 | |
Murine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title