Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-28B/IFN-lambda 3 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB5259
This item is not returnable.
View return policy
Description
IL-28B/IFN-lambda 3 Monoclonal specifically detects IL-28B/IFN-lambda 3 in Human samples. It is validated for Western Blot.Specifications
IL-28B/IFN-lambda 3 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
AAN28264 | |
IFNL3 | |
Chinese hamster ovary cell line CHO-derived recombinant human IL-28B/IFN-lambda 3 Arg30-Val200 (Lys74Arg) Accession # AAN28264 | |
100 μg | |
Primary | |
Detects human IL-28B/IFN-lambda 3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-28A, rhIL-29, or recombinant mouse IL-28 is observed. | |
Human | |
Purified |
Western Blot | |
567143 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
interleukin-28B, IFN-lambda 3, IL28B, IL-28B, IL28C, interferon, lambda 3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
282617 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction