Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-29/IFN-lambda 1 Mouse anti-Human, Clone: 247801, R&D Systems™

Mouse Monoclonal Antibody

$125.00 - $839.00

Specifications

Antigen IL-29/IFN-lambda 1
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-29/IFN-lambda 1 Gly20-Thr200 Accession # Q8IU54
Clone 247801
Applications Neutralization, ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB15981SP
View Documents
R&D Systems
MAB15981SP
25 ug
Each for $125.00
Add to cart
 
MAB15981100
View Documents
R&D Systems
MAB15981100
100 ug
Each for $387.50
Add to cart
 
MAB15981500
View Documents
R&D Systems
MAB15981500
500 ug
Each for $839.00
Add to cart
 
Description

Description

IL-29/IFN-lambda 1 Monoclonal antibody MAB15981 Specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for ELISA Capture, Neutralization
Specifications

Specifications

IL-29/IFN-lambda 1
247801
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2a
282618
RUO
Mouse myeloma cell line NS0-derived recombinant human IL-29/IFN-lambda 1 Gly20-Thr200 Accession # Q8IU54
Neutralization, ELISA
Purified
Q8IU54
Monoclonal
Primary
Detects human IL-29/IFN-lambda 1 in ELISAs. In sandwich immunoassays, 100% cross-reactivity with recombinant human (rh) IL-28B/IFN-lambda 3 and 20% with rhIL-28A is observed. No cross-reactivity or interference was observed with rhIFN-alpha A, rhIFN-alpha B2, rhIFN-alpha G, rhIFN-alpha I, or rhIFN-beta 1A.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit