Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-3 Mouse anti-Human, Clone: 4815, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523695

Catalog No. PIMA523695


Add to cart

Description

Description

In sandwich ELISAs, no significant cross-reactivity or interference with recombit human (rh) IL-1 alpha, rhIL-1 beta, recombit mouse (rm) IL-1 beta, rhIL-2, rhIL-3 R alpha, rhIL-3 R beta, rmIL-3, rhIL-4, rmIL-4, rmIL-5, rhIL-6, rmIL-6, rhIL-7, rmIL-7, rhIL-8, bovine FGF acidic, bovine FGF basic, rhG-CSF, rhGM-CSF, rmGM-CSF, rhLIF, hPDGF, porcine PDGF, human TGF- beta 1, porcine TGF- beta 1, rhTGF- beta 1, porcine TGF- beta 2, porcine TGF- beta 1.2, rhTNF-alpha, or rhTNF-beta was observed. This antibody does not neutralize the biological activity of rmIL-3. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
Specifications

Specifications

IL-3
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P08700
Interleukin-3, IL3, Multipotential colony-stimulating factor, Hematopoietic growth factor, P-cell-stimulating factor, Mast-cell growth factor, MCGF
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3562
ELISA, Neutralization
4815
Unconjugated
IL3
Lyophilized
IL3
E. coli-derived recombit human IL-3 Ala20-Phe152
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit