Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-31 Mouse anti-Human, Biotin, Clone: SB118b, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 1563008
Description
SouthernBiotech cytokine antibodies offer high sensitivity and low background for use in a variety of applications.
Specifications
IL-31 | |
Monoclonal | |
0.5mg/mL | |
Supplied in PBS/NaN3 | |
E. coli-expressed human IL-31 | |
RUO | |
Human | |
IgG2b |
ELISA | |
SB118b | |
Biotin | |
Mouse | |
0.1 mg | |
Primary | |
Store at 2-8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction