Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-33, APC, clone: 390412, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523609

Catalog No. PIMA523609


Add to cart

Description

Description

Interleukin-33 (IL-33) is a recently identified member of the IL-1 family of cytokines whose other members include IL-1-a-b, IL-1Ra and IL-18. Its receptor has been shown to be ST2, an IL-1 receptor family member that also acts as a negative regulator of TLR-IL-1R signaling and IL-1R accessory protein (IL-1RAcP). Receptor binding of IL-33 activates NF-kappa-B and MAP kinases and induces the expression of TH2-associated cytokines such as IL-4, IL-5 and IL-6. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine. Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. Despite its predicted molecular weight, IL-33 will often run at higher molecular weight in SDS-PAGE.

In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-33 is observed.
Specifications

Specifications

IL-33
Monoclonal
APC
IL33
Liquid
IL33
E. coli-derived recombinant human IL-33 Ser112-Thr270
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
390412
Buffered saline solution with BSA and 0.09% sodium azide
O95760
C9orf26, DVS27, IL1F11, NF-HEV, NFEHEV, RP11-575C20.2, DVS27-related protein, IL-1F11, interleukin-1 family member 11, interleukin-33, nuclear factor for high endothelial venules, nuclear factor from high endothelial venules
Rat
IgG2B
100 Tests
Store at 4°C, store in the dark, do not freeze
Primary
90865
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit