Learn More
IL-33, PE, clone: 390412, Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA523673
Description
Interleukin-33 (IL-33) is a recently identified member of the IL-1 family of cytokines whose other members include IL-1-a-b, IL-1Ra and IL-18. Its receptor has been shown to be ST2, an IL-1 receptor family member that also acts as a negative regulator of TLR-IL-1R signaling and IL-1R accessory protein (IL-1RAcP). Receptor binding of IL-33 activates NF-kappa-B and MAP kinases and induces the expression of TH2-associated cytokines such as IL-4, IL-5 and IL-6. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine. Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. Despite its predicted molecular weight, IL-33 will often run at higher molecular weight in SDS-PAGE.
In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-33 is observed.Specifications
IL-33 | |
Monoclonal | |
PE | |
IL33 | |
C9orf26, DVS27, IL1F11, NF-HEV, NFEHEV, RP11-575C20.2, DVS27-related protein, IL-1F11, interleukin-1 family member 11, interleukin-33, nuclear factor for high endothelial venules, nuclear factor from high endothelial venules | |
Rat | |
Protein A/G | |
RUO | |
90865 | |
Store at 4°C, store in the dark, do not freeze | |
Liquid |
Flow Cytometry | |
390412 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
O95760 | |
IL33 | |
E. coli-derived recombinant human IL-33 Ser112-Thr270 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.