Learn More
IL-33R ST2 (IL1RL1) Rabbit anti-Human, Chimpanzee, Primate, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061116
Catalog No. 06-111-6MI
Add to cart
Description
Description
Anti-IL-33R ST2 (IL1RL1) Antibody is an antibody against IL-33R ST2 (IL1RL1) for use in western blot, immunohistochemistry (paraffin).
Specifications
Specifications
IL-33R ST2 (IL1RL1)
Unconjugated
Affinity Purified
Rabbit
Affinity Purified
2-8°C for 12 months
Polyclonal
Chimpanzee, Human, Primate
Immunohistochemistry (Paraffin), Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
T1; DER4; IL1RL1; ST2; FIT1; ST2L; ST2V
KLH-conjugated linear peptide corresponding to the extracellular domain of human IL-33R ST2.
100μg
Primary
NP_057316