Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Mouse anti-Rat, Clone: 154101, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen IL-4
Immunogen E. coli-derived recombinant rat IL-4 Cys25-Ser147 Accession # P20096.2
Clone 154101
Applications Western Blot, Neutralization
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB5041SP
View Documents
R&D Systems
MAB5041SP
25 ug
Each for $119.60
Add to cart
 
MAB5041100
View Documents
R&D Systems
MAB5041100
100 ug
Each for $377.00
Add to cart
 
MAB5041500
View Documents
R&D Systems
MAB5041500
500 ug
Each for $839.00
Add to cart
 
Description

Description

IL-4 Monoclonal antibody MAB5041 Specifically detects IL-4 in Rat samples. It is validated for Neutralization, Western Blotting
Specifications

Specifications

IL-4
154101
Unconjugated
P20096.2
Monoclonal
Primary
Detects recombinant rat IL-4 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human IL-4, recombinant mouse IL-4, recombinant feline IL-4, recombinant canine IL-4, recombinant porcine IL-4, recombinant cotton rat IL-4, or recombinant mouse IL-13 is observed.
Protein A or G purified from hybridoma culture supernatant
E. coli-derived recombinant rat IL-4 Cys25-Ser147 Accession # P20096.2
Western Blot, Neutralization
Purified
Mouse
IgG1
3565
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit