Learn More
IL-4R (CD124) Rabbit anti-Human, Chimpanzee, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061084
Catalog No. 06-108-4MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This Anti-IL-4R (CD124) Antibody, cytoplasmic is validated for use in ELISA, immunocytochemistry, immunohistochemistry (paraffin), FC for the detection of IL-4R (CD124).
Specifications
Specifications
IL-4R (CD124)
Unconjugated
Affinity Purified
Rabbit
Affinity Purfied
2-8°C for 12 months
Polyclonal
Chimpanzee, Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Purified rabbit polyclonal in PBS with 0.09% sodium azide.
IL4R; CD124; IL-4R-alpha; IL4RA
KLH-conjugated linear peptide corresponding to human IL-4R at and around the cytoplasmic domain.
100μg
Primary
NP_000409