Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-5 Mouse anti-Rat, Clone: 299298, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524011

Catalog No. PIMA524011


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
Specifications

Specifications

IL-5
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q08125
B cell differentiation factor I, B-cell differentiation factor I, T-cell replacing factor, eosinophil differentiation factor, interleukin-5
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
24497
Immunocytochemistry, Immunofluorescence, Neutralization
299298
Unconjugated
Il5
Lyophilized
IL5
Sf 21-derived recombit rat IL-5 Met20-Val132
Protein A/G
RUO
Antibody
Monoclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit