Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA120673

Catalog No. PIPA120673


Add to cart

Description

Description

PA1-20673 detects CXCR2 in Human samples. PA1-20673 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA1-20673 immunogen corresponds to Synthetic peptide (aa sequence is considered to be commercially sensitive).

CXCR2 is a receptor for interleukin-8, which is a powerful neutrophil chemotactic factor. It is a member of the GPCR family. Binding of IL-8 to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activate a phosphatidylinositol-calcium second messenger system. This receptor binds to IL-8 with a high affinity and to GRO/MGSA and NAP-2 also with a high affinity. It has been reported to be expressed in a wide variety of tissues. ESTs have been isolated from human placenta and thymus libraries.
Specifications

Specifications

CXCR2
Polyclonal
Unconjugated
CXCR2
Liquid
CXCR2
Synthetic peptide (aa sequence is considered to be commercially sensitive)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide; pH 7.4
P25025
CXCR2, CDw128b, CD182 antigen, CMKAR2, CXCR-2, CD182, CXC-R2, IL-8R B, Il-8 receptor b, IL-8 receptor type 2, Il-8rb, IL8R2, IL8RB, IL-8 receptor beta
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3579
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit