Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9 Mouse anti-Human, APC, Clone: 623153, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523679

Catalog No. PIMA523679


Add to cart

Description

Description

Interleukin 9 (IL9) is a cytokine that regulates a variety of hematopoietic cells. It is secreted by T-cells, and supports IL-2 independent, and IL-4 independent growth of helper T-cells. It is also a positive regulator of cell proliferation, and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates signal transducer and activator (STAT) proteins and results in IRS1 and IRS2/PI3K and MEK/ERK signaling cascades.
Specifications

Specifications

IL-9
Monoclonal
APC
IL9
Liquid
IL9
Sf 21-derived recombit human IL-9 Gln19-Ile144
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
623153
Buffered saline solution with BSA and 0.09% sodium azide
P15248
Interleukin-9
Mouse
IgG2B
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3578
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit