Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9 Rat anti-Mouse, Clone: 222604, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523789

Catalog No. PIMA523789


Add to cart

Description

Description

In Western blots, no cross-reactivity with recombit human IL-9 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Interleukin 9 (IL9) is a cytokine that regulates a variety of hematopoietic cells. It is secreted by T-cells, and supports IL-2 independent, and IL-4 independent growth of helper T-cells. It is also a positive regulator of cell proliferation, and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates signal transducer and activator (STAT) proteins and results in IRS1 and IRS2/PI3K and MEK/ERK signaling cascades.
Specifications

Specifications

IL-9
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P15247
Interleukin-9
Rat
IgG2a
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
16198
Flow Cytometry, Western Blot
222604
Unconjugated
Il9
Lyophilized
IL9
Sf 21-derived recombit mouse IL-9 Gln19-Pro144
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit