Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
interleukin 13 receptor, alpha 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human IL13RA2 protein.
Supplier: Abnova Corporation H00003598D01P
Description
The protein encoded by this gene is closely related to Il13RA1, a subuint of the interleukin 13 receptor complex. This protein binds IL13 with high affinity, but lacks cytoplasmic domain, and does not appear to function as a signal mediator. It is reported to play a role in the internalization of IL13. [provided by RefSeq
Sequence: MAFVCLAIGCLYTFLISTTFGCTSSSDTEIKVNPPQDFEIVDPGYLGYLYLQWQPPLSLDHFKECTVEYELKYRNIGSETWKTIITKNLHYKDGFDLNKGIEAKIHTLLPWQCTNGSEVQSSWAETTYWISPQGIPETKVQDMDCVYYNWQYLLCSWKPGIGVLLDTNYNLFYWYEGLDHALQCVDYIKADGQNIGCRFPYLEASDYKDFYICVNGSSENKPIRSSYFTFQLQNIVKPLPPVYLTFTRESSCEIKLKWSIPLGPIPARCFDYEIEIREDDTTLVTATVENETYTLKTTNETRQLCFVVRSKVNIYCSDDGIWSEWSDKQCWEGEDLSKKTLLRFWLPFGFILILVIFVTGLLLRKPNTYPKMIPEFFCDTSpecifications
interleukin 13 receptor, alpha 2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human IL13RA2 protein. | |
IL13RA2 | |
CD213A2/IL-13R/IL13BP | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Flow Cytometry, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000640.2 | |
IL13RA2 | |
IL13RA2 (NP_000631.1, 1 a.a. ∼ 380 a.a) full-length human protein. | |
100 μg | |
Membrane Receptors | |
3598 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction